Learn More
MilliporeSigma™ Supelco™ Purge/Trap I Vocarb™ 4000
Specific configurations available for a variety of instruments
$179.82 - $436.77
Specifications
Type Purge/Trap I
For Use With (Application) Industrial Hygiene
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
111004293
View Documents Promotion Details
MilliporeSigma™ Supelco™
24938
OI Analytical Eclipse 4660, 4560
Each for $436.77
Only null left
111004526
View Documents Promotion Details
MilliporeSigma™ Supelco™
20308
Tekmar Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000, HT-3
Each for $204.61
Only null left
111004592
View Documents Promotion Details
MilliporeSigma™ Supelco™
24918
Tekmar 3000, 3100
Each for $179.82
Only null left
Description
Description
  • Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials
  • A single tube can trap a broad range of polar and nonpolar, high and low molecular weight compounds
  • Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition
  • During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds
  • During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents
Specifications
Specifications
Purge/Trap I
Industrial Hygiene
Videos
SDS
Documents
Documents
Vocarb is a registered trademark of Siemens Industry, Inc.