missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Punicalagin, ≥85% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 65995-63-3 |
| Physical Form | Neat |
| Linear Formula | C48H28O30 |
| MDL Number | MFCD09838017 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 1084.72 |
| Percent Purity | ≥85% (HPLC) |
| Grade | Analytical Standard |