missing translation for 'onlineSavingsMsg'
Learn More

Pullulan

Catalog No. aaj6696106
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6696106 5 g
AAJ6696103 1 g
AAJ6696109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6696106 Supplier Thermo Scientific Chemicals Supplier No. J6696106
Only null left

CAS: 9057-02-7 | (C18H30O15)AHO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9057-02-7

Specifications

Color White
Molecular Formula (C18H30O15)AHO
Quantity 5 g
MDL Number MFCD00081940
InChI Key GFZFEWWPMNSVBS-WVZDODFGSA-N
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC)CO)CO)O)O)O
IUPAC Name (2R,4S,5S)-2-[(3S,4R,6R)-6-[(3S,4R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(methoxymethyl)oxane-3,4,5-triol
PubChem CID 131636581
Physical Form Powder
Chemical Name or Material Pullulan
EINECSNumber 232-945-1
RTECSNumber UO5470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only