missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Pulegone 85.0+%, TCI America™
Chemical Identifiers
| CAS | 89-82-7 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.237 |
| MDL Number | MFCD00063000 |
| InChI Key | NZGWDASTMWDZIW-QMMMGPOBSA-N |
| Synonym | --pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one |
| PubChem CID | 638012 |
| ChEBI | CHEBI:81226 |
| IUPAC Name | (5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one |
| SMILES | CC1CCC(=C(C)C)C(=O)C1 |
Chemical Identifiers
| C10H16O | |
| MFCD00063000 | |
| --pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one | |
| CHEBI:81226 | |
| CC1CCC(=C(C)C)C(=O)C1 |
Specifications
| 89-82-7 | |
| 217°C | |
| MFCD00063000 | |
| NZGWDASTMWDZIW-QMMMGPOBSA-N | |
| (5S)-5-methyl-2-propan-2-ylidenecyclohexan-1-one | |
| 638012 | |
| 152.24 | |
| Liquid |
| Yellow | |
| C10H16O | |
| --pulegone, s---pulegone, pulegone,-, unii-hl7z89m60h, cyclohexanone, 5-methyl-2-1-methylethylidene-, s, s-p-menth-4 8-en-3-one, s-2-isopropylidene-5-methylcyclohexanone, 5s-5-methyl-2-propan-2-ylidenecyclohexan-1-one | |
| CC1CCC(=C(C)C)C(=O)C1 | |
| 152.237 | |
| CHEBI:81226 | |
| ≥85.0% (GC) | |
| (+)-Pulegone |
Safety and Handling
EINECSNumber : (3)-2400
RTECSNumber : OT0261000
TSCA : Yes