missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Puerarin | |
| 187°C | |
| C21H20O9 | |
| 1 g | |
| HKEAFJYKMMKDOR-VPRICQMDSA-N | |
| 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one | |
| 53384442 | |
| ≥98.0% (HPLC,T) |
| 3681-99-0 | |
| White-Yellow | |
| MFCD00063399 | |
| 8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone | |
| OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1 | |
| 416.38 | |
| 416.38 | |
| Crystalline Powder |
Chemical Identifiers
| 3681-99-0 | |
| 416.38 | |
| HKEAFJYKMMKDOR-VPRICQMDSA-N | |
| 53384442 | |
| OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1 |
| C21H20O9 | |
| MFCD00063399 | |
| 8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone | |
| 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one |
Safety and Handling
RTECSNumber : UO5216000
TSCA : No
Recommended Storage : Refrigerator