missing translation for 'onlineSavingsMsg'
Learn More

Puerarin 98.0+%, TCI America™

Catalog No. P1886200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1886200MG 200 mg
P18861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1886200MG Supplier TCI America Supplier No. P1886200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3681-99-0
Molecular Formula C21H20O9
Molecular Weight (g/mol) 416.38
MDL Number MFCD00063399
InChI Key HKEAFJYKMMKDOR-VPRICQMDSA-N
Synonym 8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone
PubChem CID 53384442
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 200 mg
Formula Weight 416.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Puerarin
RTECSNumber UO5216000
TSCA No
Recommended Storage Refrigerator