Learn More
Puerarin, 98%
CAS: 3681-99-0 | C21H20O9 | 416.38 g/mol
Supplier: Thermo Scientific Chemicals J6652806
Description
Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
Specifications
| Puerarin | |
| 187°C to 189°C | |
| C21H20O9 | |
| 5 g | |
| 8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone | |
| HKEAFJYKMMKDOR-VPRICQMDSA-N | |
| 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one | |
| 53384442 | |
| 98% |
| 3681-99-0 | |
| White | |
| MFCD00063399 | |
| 64198 | |
| Soluble in DMSO or DMF; Slightly soluble in water or ethanol | |
| OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1 | |
| 416.38 | |
| 416.38 | |
| Powder |
Chemical Identifiers
| 3681-99-0 | |
| 416.38 | |
| HKEAFJYKMMKDOR-VPRICQMDSA-N | |
| 53384442 | |
| OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1 |
| C21H20O9 | |
| MFCD00063399 | |
| 8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone | |
| 7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one |
Safety and Handling
RTECSNumber : UO5216000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only