missing translation for 'onlineSavingsMsg'
Learn More

Puerarin, 80% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014051
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014051 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014051 Supplier Merck Emd Millipore Supplier No. P5555100MG

missing translation for 'validMainframeMsg'

Specifications

CAS 3681-99-0
Physical Form Neat
Linear Formula C21H20O9
MDL Number MFCD00076007
Synonym 8-(beta-D-Glucopyranosyl-7-hydroxy-3- (4-hydroxyphenyl)-4 H-1-benzopyran-4-one; Kakonein
Type Analytical Standard
Quantity 100 mg
Formula Weight 416.38
Percent Purity 80% (HPLC)
Grade Analytical Standard