missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRO Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149791
Description
PTPRO Polyclonal Antibody for Western Blot, ICC/IF, IHC
Glomerular epithelial protein 1 (GLEPP1), also designated protein tyrosine phosphatase U2 (PTPase U2) or receptor-type tyrosine-protein phosphatase O (PTPRO), belongs to the protein-tyrosine phosphatase family. GLEPP1 is a Type I membrane protein containing 8 fibronectin type-III domains and 1 tyrosine-protein phosphatase domain. GLEPP1 is expressed primarily in the glomeruli of the kidney, but is also detected in placenta, lung and brain.Specifications
| PTPRO | |
| Polyclonal | |
| Unconjugated | |
| PTPRO | |
| D28; FMI; GLEPP1; Glomerular epithelial protein 1; hPTP-J; NPHS6; osteoclastic transmembrane protein-tyrosine phosphatase; PCP2; PCP-2; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; protein tyrosine phosphatase U2; protein tyrosine phosphatase, non-receptor type 15; protein tyrosine phosphatase, receptor type O; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP pi; PTPase U2; PTPBK; PTP-J; Ptpn15; PTPoc; PTP-OC; PTPphi; Ptpro; PTP-RO; PTPROT; PTPRU; PTPU2; PTP-U2; receptor protein tyrosine phosphatase RO; receptor-type protein tyrosine phosphatase D30; receptor-type tyrosine-protein phosphatase O; RPTP-BK; R-PTP-O; R-PTP-psi; R-PTP-U | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19277, 50677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| E9Q612 | |
| PTPRO | |
| Recombinant Protein Tyrosine Phosphatase Receptor Type O (PTPRO) (Phe948-Met1205). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody |