missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTPRB Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5149853

Catalog No. PA5149853


Only null left

Description

Description

PTPRB Polyclonal Antibody for IHC (P), Western Blot

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and one intracytoplasmic catalytic domain, thus belongs to receptor type PTP. The extracellular region of this PTP is composed of multiple fibronectin type_III repeats, which was shown to interact with neuronal receptor and cell adhesion molecules, such as contactin and tenascin C. This protein was also found to interact with sodium channels, and thus may regulate sodium channels by altering tyrosine phosphorylation status. The functions of the interaction partners of this protein implicate the roles of this PTP in cell adhesion, neurite growth, and neuronal differentiation. Alternate transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

PTPRB
Polyclonal
Unconjugated
PTPRB
3230402H02Rik; C130094E24; HPTPB; HPTP-BETA; protein tyrosine phosphatase, receptor type B; protein tyrosine phosphatase, receptor type, B; protein tyrosine phosphatase, receptor type, beta polypeptide; protein-tyrosine phosphatase beta; PTPB; Ptprb; Ptpz; receptor-type tyrosine-protein phosphatase beta; Rptpb; R-PTP-BETA; vascular endothelial protein tyrosine phosphatase; VEPTP; VE-PTP
Rabbit
Protein A
RUO
5787
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P23467
PTPRB
Recombinant Protein Tyrosine Phosphatase Receptor Type B (PTPRB) (Ala1655-Asp1918).
200 μL
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions