missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTIO (=2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) 98.0+%, TCI America™
[Stable free radical reagent for the simultaneous determination of NO and NO2Â in the atmosphere]
$279.60 - $838.83
Chemical Identifiers
| CAS | 18390-00-6 |
|---|---|
| Molecular Formula | C13H17N2O2 |
| MDL Number | MFCD00059929 |
| Synonym | 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl |
Chemical Identifiers
| 18390-00-6 | |
| C13H17N2O2 | |
| MFCD00059929 | |
| 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl |
Specifications
| 18390-00-6 | |
| C13H17N2O2 | |
| 1 g | |
| 233.29 | |
| Crystalline Powder |
| Blue | |
| MFCD00059929 | |
| 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl | |
| ≥98.0% (T) | |
| PTIO (=2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) [Stable free radical reagent f |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator