accessibility menu, dialog, popup

UserName

CAS RN 18390-00-6

CH3H3CNONOH3CH3C

CAS RN 18390-00-6

IUPAC Name: (4,4,5,5-tetramethyl-3-oxido-2-phenyl-4,5-dihydro-1H-imidazol-3-ium-1-yl)oxidanyl
Synonyms: 4,4,5,5-tetramethyl-3-oxido-2-phenylimidazol-3-ium-1-yloxidanyl
Molecular Weight (g/mol): 233.29
Molecular Formula: C13H17N2O2
InChi Key: DYUUGILMVYJEHY-UHFFFAOYSA-N
SMILES: CC1(C)N([O])C(C2=CC=CC=C2)=[N+]([O-])C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results