missing translation for 'onlineSavingsMsg'
Learn More

PTIO (=2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) 98.0+%, TCI America™

Catalog No. A54405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54405G 5 g
A54401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A54405G Supplier TCI America Supplier No. A54405G
Only null left

[Stable free radical reagent for the simultaneous determination of NO and NO2 in the atmosphere]

Chemical Identifiers

CAS 18390-00-6
Molecular Formula C13H17N2O2
MDL Number MFCD00059929
Synonym 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl

Specifications

Color Blue
Quantity 5 g
Formula Weight 233.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material PTIO (=2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) [Stable free radical reagent f
TSCA No
Recommended Storage Refrigerator