missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTGER4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534125
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534125 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534125 Supplier Invitrogen™ Supplier No. PA534125

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Chimpanzee, Gorilla, Marmoset (93%) Gibbon, Monkey (86%).

The protein encoded by this gene is a member of the G-protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2. This receptor can activate T-cell factor signaling. It has been shown to mediate PGE2 induced expression of early growth response 1, regulate the level and stability of cyclooxygenase-2 mRNA, and lead to the phosphorylation of glycogen synthase kinase-3. Knockout studies in mice suggest that this receptor may be involved in the neonatal adaptation of circulatory system, osteoporosis, as well as initiation of skin immune responses.

Specifications

Antigen PTGER4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PTGER4
Gene Accession No. P35408
Gene Alias EP1; EP4; EP4R; MGC126583; PGE receptor EP4 subtype; PGE receptor, EP4 subtype; PGE2 receptor EP4 subtype; prostaglandin E receptor 4; prostaglandin E receptor 4 (subtype EP4); prostaglandin E2 receptor EP4 subtype; prostaglandin E2 receptor type 4; prostanoid EP4 receptor; Ptger; PTGER2; Ptger4; Ptgerep4
Gene Symbols PTGER4
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from N-terminal extracellular domain of human PTGER4 / EP4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less