Learn More
MilliporeSigma™ Supelco™ PTFE Body Two-Way Valve
Catalog No. 111010745
Change view
Click to view available options
For Use With: Hamilton PTFE Luer Lock (TLL) and Other Luer Lock Syringes
Catalog No.
111010745 For Use With: Hamilton PTFE Luer Lock (TLL) and Other Luer Lock Syringes
1 options
Catalog No. 111010745 Supplier MilliporeSigma™ Supelco™ Supplier No. 20926
This miniature, two-way valve from Hamilton provides flexibility, inertness, and small size for collecting gas samples and transporting or storing them in the original sampling syringe.
Fluid path is made entirely of PTFE to ensure inertness. Can be autoclaved for applications requiring sterility. 100 psig maximum pressure. Fits Hamilton PTFE Luer lock (TLL) and other Luer lock syringes.
  • Lever-actuated on/off valve for storing and transporting samples.
  • KEL-F hub needles attach to the valve
  • Bore I.D. 0.39 in.
Specifications
Material PTFE (Valve Body)
For Use With (Equipment) Hamilton PTFE Luer Lock (TLL) and Other Luer Lock Syringes