missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Psoralen | |
| 163°C | |
| MFCD00010520 | |
| psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin | |
| O=C1OC2=CC3=C(C=CO3)C=C2C=C1 | |
| 186.17 | |
| CHEBI:27616 | |
| ≥99.0% (GC) |
| 66-97-7 | |
| C11H6O3 | |
| 100 mg | |
| ZCCUUQDIBDJBTK-UHFFFAOYSA-N | |
| 7H-furo[3,2-g]chromen-7-one | |
| 6199 | |
| 186.17 |
Chemical Identifiers
| 66-97-7 | |
| 186.17 | |
| ZCCUUQDIBDJBTK-UHFFFAOYSA-N | |
| 6199 | |
| 7H-furo[3,2-g]chromen-7-one |
| C11H6O3 | |
| MFCD00010520 | |
| psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin | |
| CHEBI:27616 | |
| O=C1OC2=CC3=C(C=CO3)C=C2C=C1 |
Safety and Handling
RTECSNumber : LV0944000
TSCA : No
Recommended Storage : Refrigerator