missing translation for 'onlineSavingsMsg'
Learn More

Psoralen 99.0+%, TCI America™

Catalog No. p-7135393
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P207720MG 20 mg
P2077100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P207720MG Supplier TCI America Supplier No. P207720MG

Chemical Identifiers

CAS 66-97-7
Molecular Formula C11H6O3
Molecular Weight (g/mol) 186.17
MDL Number MFCD00010520
InChI Key ZCCUUQDIBDJBTK-UHFFFAOYSA-N
Synonym psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
PubChem CID 6199
ChEBI CHEBI:27616
IUPAC Name 7H-furo[3,2-g]chromen-7-one
SMILES O=C1OC2=CC3=C(C=CO3)C=C2C=C1

Specifications

Melting Point 163°C
Quantity 20 mg
Formula Weight 186.17
Percent Purity ≥99.0% (GC)
Chemical Name or Material Psoralen
RTECSNumber LV0944000
TSCA No
Recommended Storage Refrigerator