missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSME1 Monoclonal Antibody (KAA124_8B6), Invitrogen™
Mouse Monoclonal Antibody
$1693.48 - $2847.38
Specifications
| Antigen | PSME1 |
|---|---|
| Clone | KAA124_8B6 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunoprecipitation, Western Blot, Western Blot |
| Classification | Monoclonal |
Description
PSME1 Monoclonal antibody specifically detects PSME1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| PSME1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 11S regulator complex subunit alpha; 29-kD MCP activator subunit; activator of multicatalytic protease subunit 1; AW413925; epididymis secretory sperm binding protein Li 129m; HEL-S-129m; I79_008549; IFI5111; IGUP I-5111; interferon gamma up-regulated I-5111 protein; interferon-gamma IEF SSP 5111; interferon-gamma-inducible protein 5111; MGC8628; OTTHUMP00000164702; PA28 alpha subunit; PA28a; PA28alpha; protease (prosome, macropain) 28 subunit, alpha; proteasome (prosome, macropain) 28 subunit, alpha; proteasome (prosome, macropain) activator subunit 1; proteasome (prosome, macropain) activator subunit 1 (PA28 alpha); proteasome activator 28 subunit alpha; Proteasome activator complex subunit 1; proteasome activator subunit 1; Psme1; REGalpha; REG-alpha | |
| Recombinant human PSME1 (immunogen range is 1-249). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PSME1 |
| KAA124_8B6 | |
| ELISA, Immunoprecipitation, Western Blot, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q06323 | |
| 5720 | |
| IgG1 | |
| Protein A | |
| Antibody |