missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC1 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, D. melanogaster and Chicken
The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.
Specifications
Specifications
| Antigen | PSMC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | PSMC1 |
| Gene Accession No. | P62191, P62192 |
| Gene Alias | MGC24583, MGC8541, P26S4, S4, p56 |
| Gene Symbols | Psmc1 |
| Host Species | Rabbit |
| Show More |