Learn More
Invitrogen™ PSMB7 Monoclonal Antibody (OTI1E1)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA525896
Catalog No. PIMA525896
Only null left
Explore available promotions
Description
Description
PSMB7 Monoclonal Antibody for Western Blot, IHC (P)
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit in the proteasome. Expression of this catalytic subunit is downregulated by gamma interferon and proteolytic processing is required to generate a mature subunit. This subunit is not present in the immunoproteasome and is replaced by catalytic subunit 2i (proteasome beta 10 subunit).
Specifications
Specifications
PSMB7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P70195, Q99436
Psmb7
Human recombinant protein fragment corresponding to amino acids 58-277 of PSMB7 produced in E.coli.
100 μL
Primary
Human, Mouse, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1E1
Unconjugated
Psmb7
AU020723; beta-type proteasome 7 subunit; EGK_07890; hypothetical protein LOC511207; macropain chain Z; MC14; Mmc14; Multicatalytic endopeptidase complex chain Z; proteasome (prosome, macropain) subunit, beta type 7; proteasome (prosome, macropain) subunit, beta type, 7; proteasome 20S subunit beta 7; proteasome beta 7 subunit; proteasome catalytic subunit 2; proteasome subunit alpha; proteasome subunit beta 7; proteasome subunit beta type-7; proteasome subunit beta type-7 proprotein; proteasome subunit Z; protein serine kinase c17; Psmb7; RP11-101K10.7; Z
Mouse
Affinity Chromatography
RUO
19177, 5695
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions