missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ pSIVA Apoptosis Detection Flow Cytometry Kit
Specifications
Specifications
| For Use With (Application) | Flow Cytometry, Flow (Cell Surface) |
| Product Type | Flow Assay Kit |
| Content And Storage | pSIVA-IANBD, Propidium Iodide Staining Solution, 10X PBS Buffer, 10X Binding Buffer. Storage is content dependent. |
| Conjugate | IANBD |
| Product Line | Novus |
| Quantity | 25 Tests |