missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PSG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pa5149892
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5149892 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5149892 Supplier Invitrogen™ Supplier No. PA5149892
Only null left

Rabbit Polyclonal Antibody

PSG2 Polyclonal Antibody for Western Blot, ICC/IF, IHC

The human pregnancy-specific glycoproteins (PSGs) are a family of proteins that are synthesized in large amounts by placental trophoblasts and released into the maternal circulation during pregnancy. Molecular cloning and analysis of several PSG genes has indicated that the PSGs form a subgroup of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily of genes. Members of the CEA family consist of a single N domain, with structural similarity to the immunoglobulin variable domains, followed by a variable number of immunoglobulin constant-like A and/or B domains. Most PSGs have an arg-gly-asp (RGD) motif, which has been shown to function as an adhesion recognition signal for several integrins, in the N-terminal domain (summary by Teglund et al., 1994 [PubMed 7851896]). For additional general information about the PSG gene family, see PSG1 (MIM 176390).

Specifications

Antigen PSG2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PSG2
Gene Accession No. P11465
Gene Alias carcinoembryonic antigen SG8; CEA; pregnancy specific beta-1-glycoprotein 2; pregnancy-specific beta-1 glycoprotein E; Pregnancy-specific beta-1-glycoprotein 2; pregnancy-specific beta-1-glycoprotein 7; pregnancy-specific beta-1-glycoprotein-2; pregnancy-specific glycoprotein 2; PS-beta-E; PS-beta-G-2; PSBG2; PSBG-2; PSG1; PSG2
Gene Symbols PSG2
Host Species Rabbit
Immunogen Recombinant Pregnancy Specific Beta-1-Glycoprotein 2 (PSG2) (Gln35-Thr335).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5670
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less