missing translation for 'onlineSavingsMsg'
Learn More

Pseudothiohydantoin, 97%

CAS: 556-90-1 | C3H4N2OS | 116.138 g/mol

Supplier:  Thermo Scientific Chemicals B2146114

 View more versions of this product

Catalog No. AAB2146114

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pseudothiohydantoin
∼240°C (decomposition)
MFCD00003186
pseudothiohydantoin, 2-aminothiazol-4 5h-one, 2-imino-4-thiazolidinone, 4 5h-thiazolone, 2-amino, usaf dm-1, 2-imino-4-thiazolidone, 2-amino-2-thiazolin-4-one, usaf be-4-5, 2-amino-4-oxo-thiazole, 2-imino-thiazolidin-4-one
C1C(=O)N=C(S1)N
116.138
116.14
556-90-1
C3H4N2OS
25 g
HYMJHROUVPWYNQ-UHFFFAOYSA-N
2-amino-1,3-thiazol-4-one
11175
97%

Chemical Identifiers

556-90-1
116.138
HYMJHROUVPWYNQ-UHFFFAOYSA-N
11175
C1C(=O)N=C(S1)N
C3H4N2OS
MFCD00003186
pseudothiohydantoin, 2-aminothiazol-4 5h-one, 2-imino-4-thiazolidinone, 4 5h-thiazolone, 2-amino, usaf dm-1, 2-imino-4-thiazolidone, 2-amino-2-thiazolin-4-one, usaf be-4-5, 2-amino-4-oxo-thiazole, 2-imino-thiazolidin-4-one
2-amino-1,3-thiazol-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-145-6

RTECSNumber : XJ6276000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only