CAS RN 556-90-1
IUPAC Name:
2-amino-4,5-dihydro-1,3-thiazol-4-one
Synonyms:
4(5H)-thiazolone, 2-amino-
Molecular Weight (g/mol):
116.14
Molecular Formula:
C3H4N2OS
InChi Key:
HYMJHROUVPWYNQ-UHFFFAOYSA-N
SMILES:
NC1=NC(=O)CS1
1
–
1
of
1
results
1
Promotions Available
Pseudothiohydantoin, 97%
Pseudothiohydantoin, 97%
CAS: 556-90-1 Molecular Formula: C3H4N2OS Molecular Weight (g/mol): 116.138 MDL Number: MFCD00003186 InChI Key: HYMJHROUVPWYNQ-UHFFFAOYSA-N Synonym: pseudothiohydantoin,2-aminothiazol-4 5h-one,2-imino-4-thiazolidinone,4 5h-thiazolone, 2-amino,usaf dm-1,2-imino-4-thiazolidone,2-amino-2-thiazolin-4-one,usaf be-4-5,2-amino-4-oxo-thiazole,2-imino-thiazolidin-4-one PubChem CID: 11175 IUPAC Name: 2-amino-1,3-thiazol-4-one SMILES: C1C(=O)N=C(S1)N