missing translation for 'onlineSavingsMsg'
Learn More

Pseudothiohydantoin, 97%

Catalog No. aab2146106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2146106 5 g
AAB2146114 25 g
AAB2146122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2146106 Supplier Thermo Scientific Chemicals Supplier No. B2146106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 556-90-1 | C3H4N2OS | 116.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 556-90-1
Molecular Formula C3H4N2OS
Molecular Weight (g/mol) 116.138
MDL Number MFCD00003186
InChI Key HYMJHROUVPWYNQ-UHFFFAOYSA-N
Synonym pseudothiohydantoin, 2-aminothiazol-4 5h-one, 2-imino-4-thiazolidinone, 4 5h-thiazolone, 2-amino, usaf dm-1, 2-imino-4-thiazolidone, 2-amino-2-thiazolin-4-one, usaf be-4-5, 2-amino-4-oxo-thiazole, 2-imino-thiazolidin-4-one
PubChem CID 11175
IUPAC Name 2-amino-1,3-thiazol-4-one
SMILES C1C(=O)N=C(S1)N

Specifications

Melting Point ∼240°C (decomposition)
Quantity 5 g
Formula Weight 116.14
Percent Purity 97%
Chemical Name or Material Pseudothiohydantoin
EINECSNumber 209-145-6
RTECSNumber XJ6276000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only