missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Prussian Blue, pure

Catalog No. ac215211000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC215211000 100 g Glass bottle
AC215210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC215211000 Supplier Thermo Scientific Chemicals Supplier No. 215211000
Only null left

CAS: 14038-43-8 | C18Fe7N18 | 859.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14038-43-8
Molecular Formula C18Fe7N18
Molecular Weight (g/mol) 859.239
MDL Number MFCD00135663
InChI Key DCYOBGZUOMKFPA-UHFFFAOYSA-N
Synonym C.I. 77510, Ferric ferrocyanide, Pigment Blue 27
PubChem CID 2724251
IUPAC Name iron(2+);iron(3+);octadecacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3]

Specifications

Linear Formula Fe4[Fe(CN)6]3
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in diluted acids, practically insoluble in most common organic, solvents
Infrared Spectrum Authentic
Formula Weight 859.25
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Blue
Quantity 100 g
Physical Form Powder
Chemical Name or Material Prussian Blue
Show More Show Less
EINECSNumber 237-875-5
RTECSNumber LJ8200000
TSCA TSCA

RUO – Research Use Only