accessibility menu, dialog, popup

UserName

CAS RN 14038-43-8

IUPAC Name: tetrairon(3+) tris(λ²-iron(2+)) octadecakis(iminomethanide)
Synonyms: prussian blue
Molecular Weight (g/mol): 859.24
Molecular Formula: C18Fe7N18
InChi Key: DCYOBGZUOMKFPA-UHFFFAOYSA-N
SMILES: [Fe++].[Fe++].[Fe++].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Quantity 
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results