missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRSS33 Polyclonal Antibody
PRSS33 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA5104380
Description
PRSS33 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| PRSS33 | |
| Polyclonal | |
| Unconjugated | |
| Prss33 | |
| Eos; mT6; protease, serine 33; protease, serine, 33; Prss33; Serine protease 33; serine protease EOS; tryptase 6; Tryptase-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 260429 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NF86 | |
| Prss33 | |
| A synthesized peptide derived from human PRSS33(Accession Q8NF86), corresponding to amino acid residues A17-I67. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |