missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRSS23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149834
Description
PRSS23 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| PRSS23 | |
| Polyclonal | |
| Unconjugated | |
| PRSS23 | |
| protease, serine 23; protease, serine, 23; PRSS23; Putative secreted protein Zsig13; serine protease 23; serine protease, umbilical endothelium; SIG13; SPUVE; UNQ270/PRO307; ZSIG13 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11098, 308807, 76453 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O95084, Q6AY61, Q9D6X6 | |
| PRSS23 | |
| Recombinant Protease, Serine 23 (PRSS23) (Gln20-Gly383). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |