missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRPF40A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558319 Supplier Invitrogen™ Supplier No. PA558319
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: SQTKESRWAK PKELEDLEGY QNTIVAGSLI TKSNLHAMIK AEESSKQEEC TTTSTAPVPT TEI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

FNBP3, Formin-binding protein 3, is a 957 amino acid protein encoded by the human gene PRPF40A. FNBP3 belongs to the PRPF40 family and contains five FF domains and two WW domains. Nuclear proteins harboring both WW and FF protein interaction modules bind to splicing factors as well as RNA polymerase II and may serve to link transcription with splicing. Through the WW domains, FNBP3 will bind to the Formin proline-rich regions. Also known as Pre-mRNA-processing factor 40 homolog A, FNBP3 binds to WASL/N-WASP (Neuronal Wiskott-Aldrich syndrome protein) complex and suppresses its translocation from the nucleus to the cytoplasm, thereby inhibiting its cytoplasmic function. FNBP3 is widely expressed in most tissues and is localized to the nuclear speckles.

Specifications

Antigen PRPF40A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF40A
Gene Accession No. O75400
Gene Alias 2810012K09Rik; Fas ligand-associated factor 1; Fas-ligand associated factor 1; FBP11; FBP-11; FLAF1; FLJ20585; Fnbp11; Fnbp3; formin binding protein 11; formin binding protein 3; formin-binding protein 11; Formin-binding protein 3; HIP10; HIP-10; HSPC225; Huntingtin yeast partner A; huntingtin-interacting protein 10; Huntingtin-interacting protein A; HYPA; NY-REN-6; NY-REN-6 antigen; pre-mRNA processing factor 40 homolog A; pre-mRNA-processing factor 40 homolog A; Prp40; PRP40 pre-mRNA processing factor 40 homolog A; PRP40 pre-mRNA processing factor 40 homolog A (S. cerevisiae); PRP40 pre-mRNA processing factor 40 homolog A (yeast); PRPF40A; Renal carcinoma antigen NY-REN-6
Gene Symbols PRPF40A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRPF40A. Recombinant protein control fragment (Product #RP-102820).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less