missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRP8 Recombinant Rabbit Monoclonal Antibody (23GB1995)

Catalog No. MA552469
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552469 20 μL
MA552468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552469 Supplier Invitrogen™ Supplier No. MA552469
Only null left

Rabbit Recombinant Monoclonal Antibody

PRP8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Pre-mRNA splicing occurs in 2 sequential transesterification steps. The protein encoded by this gene is a component of both U2- and U12-dependent spliceosomes, and found to be essential for the catalytic step II in pre-mRNA splicing process. It contains several WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp8 protein. This gene is a candidate gene for autosomal dominant retinitis pigmentosa.

Specifications

Antigen PRP8
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1995
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Prpf8
Gene Accession No. Q6P2Q9, Q99PV0
Gene Alias 220 kDa U5 snRNP-specific protein; apoptosis-regulated protein 1; apoptosis-regulated protein 2; AU019467; D11Bwg0410e; DBF3/PRP8; H PRPC8; HPRP8; id:ibd1257; ik:tdsubc_2a9; im:7141966; p220; precursor mRNA processing protein; pre-mRNA processing factor 8; pre-mRNA-processing-splicing factor 8; Prp8; PRP8 homolog; PRP8 pre-mRNA processing factor 8 homolog; PRPC8; prpf8; RP13; Sfprp8l; SNRNP220; spl; Splicing factor Prp8; tdsubc_2a9; U5 snRNP-specific protein; U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p; wu:fb37c02; wu:fb73e06; xx:tdsubc_2a9; zgc:56504
Gene Symbols Prpf8
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRPF8 (2250-2335AA).
Purification Method Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10594, 192159, 287530
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less