missing translation for 'onlineSavingsMsg'
Learn More

PROTOCOL™ 10% Buffered Formalin, Fisher HealthCare™

Catalog No. 23427098
Change view
Click to view available options
Quantity:
32 oz.
2.5 gal.
Packaging:
Cubitainer
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
23427098 2.5 gal. Cubitainer
23305510 32 oz. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23427098 Supplier Epredia Supplier No. 427098
Only null left

Traditional fixative, buffered to a neutral pH

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging Cubitainer
Quantity 2.5 gal.
Chemical Name or Material Fisher PROTOCOL™ 10% Buffered Formalin