missing translation for 'onlineSavingsMsg'
Learn More

PROTOCOL™ 10% Buffered Formalin, Fisher HealthCare™

Catalog No. 23305510
Change view
Click to view available options
Quantity:
32 oz.
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
23305510 32 oz. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23305510 Supplier Epredia Supplier No. 305510

Traditional fixative, buffered to a neutral pH

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging Poly Bottle
Quantity 32 oz.
Chemical Name or Material Fisher PROTOCOL™ 10% Buffered Formalin