missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ProteoMass™ LTQ/FT-Hybrid ESI Positive Mode Calibration Mix, MilliporeSigma™ Supelco™
Description
The ProteoMass™ ESI Calibration Kit for LTQ based Hybrids, positive ion mode calibration solution, enables calibration of Thermo Scientific™ hybrid instruments equipped with ESI source in positive ion mode. It covers a mass range from m/z 138 to m/z 1822, and is therefore usable for calibrations between m/z 50 and 2000.
Specifications
Specifications
| CAS | 75-05-8 |
| Product Line | ProteoMass |
| Flash Point | 5.6°C - closed cup |
| Shipping Condition | Dry Ice |
| Concentration | Varied Conc. |
| Type | MALDI-MS Calibration Mix |
| Quantity | 1 Ea. |
ProteoMass is a trademark of Sigma-Aldrich Co. LLC