missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ProteoMass™ Insulin chain B oxidized MALDI-MS Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Vial |
| CAS | 30003-72-6 |
| Product Line | ProteoMass |
| Linear Formula | C157H232N40O47S2 |
| MDL Number | MFCD00131380 |
| Synonym | Insulin Chain B Oxidized from bovine pancreas |
| Type | MALDI-MS Standard |
| Quantity | 5 x 1 Vial |
| Formula Weight | 3495.89 |
ProteoMass is a trademark of Sigma-Aldrich Co. LLC