missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ProteoMass™ Bradykinin Fragment 1-7 MALDI-MS Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Vial |
| Product Line | ProteoMass |
| MDL Number | MFCD00076262 |
| Synonym | Bradykinin fragment 1-7, mass spec standard |
| Type | MALDI-MS Standard |
| Quantity | 5 x 10 nmol Vial |
ProteoMass is a trademark of Sigma-Aldrich Co. LLC