missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ProteoExtract™ Native Membrane Protein Extraction Kit

Catalog No. 4448101KIT
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4448101KIT 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4448101KIT Supplier MilliporeSigma™ Supplier No. 4448101KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specialized Extraction Kit designed for isolation of native membrane proteins from broad range of mammalian samples

  • Isolation of native integral membrane and membrane-associated proteins, without the need for ultracentrifugation
  • Extraction is based on association of proteins with cellular membranes rather than on their hydrophobicity. Resulting samples are suitable for use in functional assays, 2-D gel electrophoresis, and other applications

Specifications

Quantity 1 kit
Sufficient For 1 kit is sufficient for up to 20 sample extractions
Format Affinity column
Kit Contents Wash buffer, Extraction buffer 1, Extraction buffer 2, Protease inhibitor cocktail, and user protocol
Content And Storage Store at+2°C to+8°C