missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Proteinase K Solution, Molecular Biology Grade, 20 mg/mL

Catalog No. 0218398891
Change view
Click to view available options
Quantity:
1 mL
10 mL
100 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0218398891 1000 mL
0218398801 1 mL
0218398810 10 mL
0218398880 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0218398891 Supplier MP Biomedicals™ Supplier No. 0218398891

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use proteinase K solution

  • Fungus-Derived Proteinase K Purified Recombinant
  • Prepared at 20 mg/mL in 10 mM Tris-HCl, 1 mM calcium acetate, pH 7.5 containing 50% glycerol
  • Molecular Biology Grade
  • Extremely effective on native proteins
  • Can be used to rapidly inactivate endogenous nucleases such as RNases and DNases
  • Sterile filtered using 0.22 μm filter
  • Can maintain its activity for a long time at room temperature and low temperature
  • Store at 2-8°C for long-term storage

Specifications

Characterization Mixtures
Concentration 20 mg/mL
For Use With (Application) Nuclease Inactivation
Formulation 20 mg/mL in 10 mM Tris-HCl, 1 mM calcium acetate, pH 7.5 containing 50% glycerol
Molecular Weight (g/mol) 29 kDa
Preparation Method Working concentrations of the enzyme range from 50 to 250 μg/mL.
Quantity 1000 mL
Storage Requirements 2 to 8°C
Water Content 0.478
Endotoxin Concentration 29 kDa
Recombinant Recombinant
Classification GHS08
Specific Activity >40 U/mg protein
Technique PCR, RNA Purification, DNA Purification, NGS, Sample Preparation
Expression System Fungus
Form Liquid
Color Yellowish-green
Purity or Quality Grade >95%
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.