missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Standard, 2 mg/vial BSA, MilliporeSigma™ Supelco™
$239.00 - $923.00
Specifications
| Physical Form | Neat |
|---|---|
| Concentration | 2 mg/vial BSA |
| Type | Analytical Standard |
| Grade | Analytical Standard |
Description
Featured Industries: Food and BeveragesSpecifications
| Neat | |
| 2 mg/vial BSA | |
| Analytical Standard | |
| Analytical Standard |