missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Protein and Peptide Molecular Weight Markers
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
$473.00 - $557.17
Specifications
| Form | Solid |
|---|---|
| Quantity | 10 Vials |
| Storage Requirements | Unopened vials should be stored at 4°C |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
|
45-000-072
|
Cytiva
17044601 |
LMW-SDS Marker Kit |
Pack of 10 Each for $557.17
|
|
|||||
|
45-000-071
|
Cytiva
17044501 |
HMW Native Marker Kit |
Pack of 10 Each for $473.00
|
|
|||||
Description
- Get clear, sharp bands for easy measurement of migration distances
- Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining
Specifications
| Solid | |
| 10 Vials | |
| Unopened vials should be stored at 4°C |