missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein and Peptide Molecular Weight Markers

Catalog No. 45000072
Change view
Click to view available options
Product Type:
HMW Native Marker Kit
LMW-SDS Marker Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45000072 LMW-SDS Marker Kit
45000071 HMW Native Marker Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000072 Supplier Cytiva Supplier No. 17044601
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.

  • Get clear, sharp bands for easy measurement of migration distances
  • Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining

Specifications

Description LMW-SDS Marker Kit
Color White
Form Solid
For Use With (Application) For MW determinations by polyacrylamide gel electrophoresis (PAGE) in homogeneous or gradient gels.
Kit Contents Phosphorylase b: 67μg, 97000 (Mr), Rabbit muscle (source), Albumin: 83μg, 66000 (Mr), Bovine Serum (source), Ovalbumin: 147μg, 45000 (Mr), Chicken Egg white (source), Carbonic Anhydrase
Molecular Weight (g/mol) Mr 14 000
Quantity 10 Vials
Storage Requirements Unopened vials should be stored at 4°C
Product Type LMW-SDS Marker Kit