Learn More
Azure Biosystems Protein-Free Blot Blocking Buffer
Protein-free blocking solution for Western blot, 5X concentrate
Supplier: Azure Biosystems AC2112
Description
- Protein-free blocking solution optimized for fluorescent Immunoblots and Western blots
- Does not contain proteins. Azure Protein-Free Blot Blocking Buffer can reduce background in Western blots where the primary antibodies tend to cross-react with protein blockers like BSA, casein or milk protein. For low-quality primary antibodies that may require protein-based blocking agents, BSA or non-fat dry milk can be dissolved directly in the Azure Protein-Free Blot Blocking Buffer and then used to dilute the antibody.
- Best used with assays where animal-based products usage is prohibited. Serves as an excellent blocking reagent for chemiluminescent Western blotting.
- Provided as a 5x solution
- Size: 200 mL
Spécifications
| Blocking Buffer, protein-free | |
| Western Blot | |
| Liquid | |
| 200 mL |