missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Protein A - Sepharose™ 4B
Description
This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.
This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
Specifications
Specifications
| Product Type | Protein A Sepharose |
| Product Line | Sepharose |
| Column Type | Affinity |
| Stationary Phase | Protein A |
| Content And Storage | This product is supplied as a 50% suspension (25 ml Protein A-Sepharose™ 4B + 25 ml PBS) Wet Ice |
| Quantity | 25 mL |
| Form | Suspension |
For Research Use Only. Not for use in diagnostic procedures.