missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Benchtop Evidence Drying Cabinet

Catalog No. 10359159
Change view
Click to view available options
Voltage:
110 V
220 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
10359159 220 V
10359158 110 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10359159 Supplier Labconco™ Supplier No. 3390001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a compact, space-saving method of securely drying evidence in a ventilated area

Lockable cabinet maintains the chain of custody. Air passes through the door inlet prefilter, circulates up through the interior, passes through the exhaust prefilter, HEPA and carbon filters and is exhausted back to the room through the top of the cabinet. Filters protect personnel from exposure to biohazards while eliminating nuisance odors.

  • Type 304 brushed stainless steel interior and epoxy-coated steel exterior
  • Stainless-steel hanging rod
  • Three stainless-steel collapsible/removable wire mesh shelves
  • Exhaust HEPA filter, 99.99% efficient on particles 0.3 micron
  • Activated organic vapor carbon filter with 7.5 lb. (3.4 kg) porous coconut shell media
  • Built-in 75 watt blower with solid state control rated for 5 amps
  • Storage area: 26.5D x 36W x12H in. (67.3 x 91.4 x 30.5cm) with door and 14-quart plastic bucket
  • Safety glass door with magnetic seal to accommodate user-supplied padlock or optional tamper-evident tag
  • Key-activated ultraviolet light provides secondary disinfection of interior between cases
  • 2 in. (50.8cm) diameter locking casters
  • Inlet and exhaust roughing prefilters with MERV 11 rating
  • Red filter system alarm light to alert operator to low airflow conditions
  • Three-position switch: fan ON, OFF, ultraviolet light ON
  • 96 in. (2.44m) three-wired cord and plug
  • Dimensions: 28D x 36W x 45.7 in.H (71.1 x 91.4 x 116.1cm)

Order Info

Optional ducting to the outside is available.

Specifications

Product Type Evidence Drying Cabinet
Depth (English) Interior 26.5 in.
Depth (Metric) 71.1 cm
Depth (English) Shelf 19 in.
Depth (English) 28 in.
Width (English) Storage Area 36 in.
Cabinet Type Benchtop evidence drying lockable cabinet
Depth (Metric) Interior 67.31 cm
Depth (Metric) Shelf 48.26 cm
Amperage 2 A
Width (English) Cabinet 36 in.
Width (Metric) 91.4 cm
Weight (Metric) 170 kg
Width (Metric) Shelf 20.3 cm
Weight (English) 375 lb.
Width (English) Shelf 8 in.
Width (English) 36 in.
Width (Metric) Cabinet 91.4cm
With Casters Yes
Plug Type Domestic
Width (Metric) Storage Area 91.44 cm
Includes UV light
Options Available UV Light
Voltage 220 V
Height (Metric) Storage Area 30.48 cm
Hertz 50/60Hz
Height (English) 45.7 in.
Height (Metric) 114.3 cm
Height (Metric) Interior 30.48 cm
Door Type Safety glass door with magnetic seal
Filter Type HEPA and Carbon filter
Height (English) Storage Area 12 in.
Material Stainless Steel
Show More Show Less