Learn More
Labconco™ Protector™Acid Storage Cabinets
Support Labconco fume hoods and safely store and vent acids and other corrosive liquids
Supplier:  Labconco™ 9905300
 View more versions of this product
Catalog No. 16-305-70
Only null left
Explore available promotions
Includes:
Vacuum-formed PVC liner tray, two rear vent connection cutouts for venting the cabinet into a fume hood, four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.
Description
Description
  • Corrosion-resistant polyethylene-lined interior
  • Manual-closing, non-locking door(s)
Specifications
Specifications
Storage Cabinet
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
18 in.
Single Left Hinged Door
55.88 cm
31.5 to 32.75 in.
800 lb.
Light Gray
22 in.
Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in.
38.55 kg
85 lb.
362.8 kg
ADA
80 to 83.18 cm
Steel
Product Suggestions
Product Suggestions