missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™Acid Storage Cabinets

Catalog No. 1630553
Change view
Click to view available options
Door Type:
Single Left Hinged Door
Single Right Hinged Door
Width (English):
18 in.
24 in.
Height (English) Base:
31.5 to 32.75 in.
35.5 to 36.75 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Type Width (English) Height (English) Base
1630553 Single Left Hinged Door 18 in. 35.5 to 36.75 in.
1630571 Single Right Hinged Door 24 in. 31.5 to 32.75 in.
1630552 Single Right Hinged Door 18 in. 35.5 to 36.75 in.
1630570 Single Left Hinged Door 18 in. 31.5 to 32.75 in.
1630572 Single Left Hinged Door 24 in. 31.5 to 32.75 in.
1630569 Single Right Hinged Door 18 in. 31.5 to 32.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 1630553 Supplier Labconco™ Supplier No. 9901600
Only null left

Includes

Vacuum-formed PVC liner tray, two rear vent connection cutouts for venting the cabinet into a fume hood, four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.

Support Labconco fume hoods and safely store and vent acids and other corrosive liquids

  • Corrosion-resistant polyethylene-lined interior
  • Manual-closing, non-locking door(s)

Specifications

Product Type Storage Cabinet
Color Light Gray
Width (English) 18 in.
Load Bearing Capacity (English) 800 lb.
Load Bearing Capacity (Metric) 362.8 kg
Weight (English) 90 lb.
Includes Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in.
Weight (Metric) 40.82 kg
Height (Metric) Base 90.17 to 93.34 cm
Door Type Single Left Hinged Door
Height (English) Base 35.5 to 36.75 in.
Depth (English) 22 in.
Depth (Metric) 55.88 cm
For Use With (Application) Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Material Steel
Show More Show Less