missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™Acid Storage Cabinets

Support Labconco fume hoods and safely store and vent acids and other corrosive liquids

Supplier:  Labconco™ 9901400

 View more versions of this product

Catalog No. 16-305-52


Only null left
Explore available promotions

Includes:

Vacuum-formed PVC liner tray, two rear vent connection cutouts for venting the cabinet into a fume hood, four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.


Description

Description

  • Corrosion-resistant polyethylene-lined interior
  • Manual-closing, non-locking door(s)
Specifications

Specifications

Storage Cabinet
Epoxy
18 in.
Single Right Hinged Door
55.8 cm
35.5 to 36.75 in.
Light Gray
22 in.
Vacuum formed PVC liner tray, 2 rear vent connection cutouts, four leveling feet, one 8 in. filler panel
40.8 kg
90 lb.
362.87 kg
800 lb.
Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions