missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Protective Overlaminate for Write-On Labels

Catalog No. 19104533
Change view
Click to view available options
Size (English):
6.5 x 4.5 in. (W x H)
7.5 x 5.5 in. (W x H)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English)
19104533 7.5 x 5.5 in. (W x H)
19104532 6.5 x 4.5 in. (W x H)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19104533 Supplier Brady™ Supplier No. 99276

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protects handwritten data on vinyl labels from chemicals and abrasion. Brady™ Protective Overlaminate for Write-On Labels helps in maintaining a safe work environment.

  • Write-On Arc Flash Labels format
  • Sheets have an acrylic layflat liner
  • Available in 25/Pk. quantity

Specifications

Adhesive Type Permanent
Color Clear
Format Sheet
Material Polyester
Product Type Protective Overlaminate for Write-On Label
Size (English) 7.5 x 5.5 in. (W x H)
Length (English) 5.5 in.
Width (English) 7.5 in.