missing translation for 'onlineSavingsMsg'
Learn More

Promega Proteasome-Glo™ 3-Substrate System

Catalog No. PRG8532
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8532 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG8532 Supplier Promega Supplier No. G8532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Proteasome-Glo™ Chymotrypsin-Like Assay
  • Proteasome-Glo™ Trypsin-Like Assay
  • Proteasome-Glo™ Caspase-Like Assay

The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in a purified enzyme preparation.

  • Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
  • Homogeneous, luminescent enzyme-based assays
  • Simple add-mix-measure protocol
  • Results in 10-30 minutes after reagent addition

Specifications

Quantity 50 mL
Storage Requirements -30°C to -10°C