missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protamine Sulfate from Salmon sperm, TCI America™
Supplier: TCI America P06755G
Specifications
| Protamine Sulfate from Salmon sperm | |
| Powder/Crystals | |
| White | |
| Salmine Sulfate |
| 53597-25-4 | |
| MFCD00132091 | |
| 4.0 to 7.0 (1% H2O soln.) | |
| 5 g |
Chemical Identifiers
| 53597-25-4 | |
| MFCD00132091 | |
| Salmine Sulfate |
Safety and Handling
RTECSNumber : VO9470000
TSCA : No